Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Psmf1
Entrez Gene ID
228769
Gene Name
proteasome (prosome, macropain) inhibitor subunit 1
Gene Aliases
AW048666, BC012260, PI31
Chromosome Location
Chr.2: 151716062 - 151744188 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK078035.1 - 1 - 2 268 69
AK083431.1 - 1 - 2 284 69

View Full Product Details