The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011340.3 | NP_035470.3 | 1 - 2 | 128 | 73 |
GenBank mRNA | AK012411.1 | - | 1 - 2 | 58 | 73 |
BC019852.1 | - | 1 - 2 | 38 | 73 | |
D50460.1 | - | 1 - 2 | 97 | 73 |