Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ube2g1
Entrez Gene ID
67128
Gene Name
ubiquitin-conjugating enzyme E2G 1
Gene Aliases
2700059C12Rik, AI256795, AU014992, AW552068, D130023C12Rik
Chromosome Location
Chr.11: 72607261 - 72686481 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025985.4 NP_080261.2 4 - 5 759 83
GenBank mRNA AK012539.1 - 4 - 5 666 83
AK013902.1 - 4 - 5 640 83
AK083029.1 - 4 - 5 702 83
AK167472.1 - 4 - 5 625 83
AK209284.1 - 1 - 2 84
BC065083.1 - 5 - 6 853 83
BC096474.1 - - 876 83

View Full Product Details