The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077696.1 | NP_001071164.1 | 11 - 12 | 1732 | 81 |
NM_001284248.1 | NP_001271177.1 | 13 - 14 | 2244 | 81 | |
NM_001284249.1 | NP_001271178.1 | 12 - 13 | 1841 | 81 | |
NM_001284250.1 | NP_001271179.1 | 11 - 12 | 1729 | 81 | |
NM_010412.3 | NP_034542.3 | 11 - 12 | 1729 | 81 | |
GenBank mRNA | AK122325.2 | - | 6 - 7 | 768 | 81 |
AK139202.1 | - | 12 - 13 | 1815 | 81 | |
AK145972.1 | - | 11 - 12 | 1695 | 81 | |
AK146483.1 | - | 11 - 12 | 1668 | 81 | |
AK155335.1 | - | 11 - 12 | 1703 | 81 | |
AK156008.1 | - | 5 - 6 | 2566 | 81 | |
AK157159.1 | - | 5 - 6 | 2566 | 81 | |
BC023309.1 | - | 2 - 3 | 81 | ||
BC049926.1 | - | 11 - 12 | 81 | ||
BC058554.1 | - | 13 - 14 | 2196 | 81 | |
BC060609.1 | - | 10 - 11 | 1523 | 81 |