The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077696.1 | NP_001071164.1 | 8 - 9 | 1266 | 100 |
NM_001284248.1 | NP_001271177.1 | 10 - 11 | 1778 | 100 | |
NM_001284249.1 | NP_001271178.1 | 9 - 10 | 1375 | 100 | |
NM_001284250.1 | NP_001271179.1 | 8 - 9 | 1263 | 100 | |
NM_010412.3 | NP_034542.3 | 8 - 9 | 1263 | 100 | |
GenBank mRNA | AF006602.2 | - | 7 - 8 | 979 | 100 |
AF207748.1 | - | 7 - 8 | 898 | 100 | |
AK122325.2 | - | 3 - 4 | 302 | 100 | |
AK139202.1 | - | 9 - 10 | 1349 | 100 | |
AK145972.1 | - | 8 - 9 | 1229 | 100 | |
AK146483.1 | - | 8 - 9 | 1202 | 100 | |
AK155335.1 | - | 8 - 9 | 1237 | 100 | |
AK200052.1 | - | 1 - 2 | 127 | 100 | |
BC049926.1 | - | 8 - 9 | 1199 | 100 | |
BC058554.1 | - | 10 - 11 | 1730 | 100 | |
BC060609.1 | - | 7 - 8 | 1057 | 100 |