Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ubtf
Entrez Gene ID
21429
Gene Name
upstream binding transcription factor, RNA polymerase I
Gene Aliases
A930005G04Rik, NOR-90, Tcfubf, UBF, UBF-1, UBF1
Chromosome Location
Chr.11: 102304563 - 102319757 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK004961.1 - 6 - 7 726 71
AK038139.1 - 1 - 2 1744 71
AK128947.1 - 6 - 7 753 71
AK158861.1 - 6 - 7 725 71
AK158935.1 - 6 - 7 684 71
BC004676.1 - 6 - 7 859 71
CD807309.1 - 3 - 4 307 71
X60831.1 - 5 - 6 593 71

View Full Product Details