Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ubtf
Entrez Gene ID
21429
Gene Name
upstream binding transcription factor, RNA polymerase I
Gene Aliases
A930005G04Rik, NOR-90, Tcfubf, UBF, UBF-1, UBF1
Chromosome Location
Chr.11: 102304563 - 102319757 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK004961.1 - 8 - 9 977 54
AK038139.1 - 4 - 5 2106 54
AK128947.1 - 8 - 9 1004 54
AK158861.1 - 8 - 9 976 54
AK158935.1 - 8 - 9 935 54
BC004676.1 - 9 - 10 1221 54
CK392577.1 - 4 - 5 408 54
X60831.1 - 8 - 9 955 54

View Full Product Details