Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Coro6
Entrez Gene ID
216961
Gene Name
coronin 6
Gene Aliases
-
Chromosome Location
Chr.11: 77462321 - 77470484 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139130.2 NP_624356.1 5 - 6 754 61
GenBank mRNA BC145575.1 - 5 - 6 832 61

View Full Product Details