Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Atxn7l3
Entrez Gene ID
217218
Gene Name
ataxin 7-like 3
Gene Aliases
E030022H21Rik
Chromosome Location
Chr.11: 102289300 - 102296643 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC059880.1 - 2 - 3 261 103

View Full Product Details