Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Atxn7l3
Entrez Gene ID
217218
Gene Name
ataxin 7-like 3
Gene Aliases
E030022H21Rik
Chromosome Location
Chr.11: 102289300 - 102296643 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098836.1 NP_001092306.1 8 - 9 866 103
NM_001098837.1 NP_001092307.1 8 - 9 845 103
GenBank mRNA AK090368.1 - 8 - 9 847 103
BC059880.1 - 2 - 3 261 160
BC130269.1 - 7 - 8 616 103
BM950535.1 - 6 - 7 423 103

View Full Product Details