The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038230.2 | NP_001033319.1 | 1 - 2 | 337 | 107 |
GenBank mRNA | AA269705.1 | - | 1 - 2 | 163 | 107 |
BC023039.1 | - | 1 - 2 | 190 | 107 | |
W61838.1 | - | 1 - 2 | 169 | 107 |