Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rprd1b
Entrez Gene ID
70470
Gene Name
regulation of nuclear pre-mRNA domain containing 1B
Gene Aliases
2610304G08Rik, 2810446G03Rik, Crept
Chromosome Location
Chr.2: 158028489 - 158078207 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291136.1 NP_001278065.1 1 - 2 551 109
GenBank mRNA AK045404.1 - 1 - 2 357 109

View Full Product Details