The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019945.2 | NP_064329.2 | 3 - 4 | 327 | 62 |
GenBank mRNA | AF077818.1 | - | 3 - 4 | 313 | 62 |
BC054524.1 | - | 3 - 4 | 327 | 62 |