The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028877.1 | NP_083153.1 | 6 - 7 | 462 | 63 |
GenBank mRNA | AK014493.1 | - | 5 - 6 | 1507 | 63 |
BC113196.1 | - | 6 - 7 | 465 | 63 | |
BC113802.1 | - | 6 - 7 | 460 | 63 | |
EF208034.1 | - | 6 - 7 | 462 | 63 |