Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zhx3
Entrez Gene ID
320799
Gene Name
zinc fingers and homeoboxes 3
Gene Aliases
1810059C13Rik, 4932418O04Rik, 9530010N21Rik, Tix1, mKIAA0395
Chromosome Location
Chr.2: 160770447 - 160872990 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177263.3 NP_796237.2 1 - 2 203 109
GenBank mRNA AK143296.1 - 1 - 2 184 109
BC058111.1 - 1 - 2 203 109

View Full Product Details