Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdh13
Entrez Gene ID
12554
Gene Name
cadherin 13
Gene Aliases
4932416G01Rik, Cdht, T-cadherin, Tcad
Chromosome Location
Chr.8: 118032249 - 119324929 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019707.4 NP_062681.2 4 - 5 580 73
GenBank mRNA AB022100.1 - 4 - 5 589 73
AK029195.1 - 4 - 5 539 73
AK032409.1 - 4 - 5 609 73
AK034995.1 - 4 - 5 560 73
AK039438.1 - 4 - 5 602 73
AK045201.1 - 4 - 5 581 73
AK048724.1 - 4 - 5 580 73
AK134649.1 - 4 - 5 539 73
BC021628.1 - 4 - 5 594 73

View Full Product Details