Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Cfap20
- Entrez Gene ID
-
14894
- Gene Name
- cilia and flagella associated protein 20
- Gene Aliases
-
2600014O15Rik, AL024127, Gtl3, T10-2A2
- Chromosome Location
-
Chr.8: 95420249 - 95434869 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC144770.1 |
- |
1 - 2 |
284 |
83 |
View Full Product Details