Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Aprt
Entrez Gene ID
11821
Gene Name
adenine phosphoribosyl transferase
Gene Aliases
C85684
Chromosome Location
Chr.8: 122574637 - 122576907 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK182364.1 - 2 - 3 162 72

View Full Product Details