The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001047436.2 | NP_001040901.1 | 2 - 3 | 390 | 74 |
NM_001286344.1 | NP_001273273.1 | 2 - 3 | 359 | 74 | |
NM_019695.3 | NP_062669.2 | 2 - 3 | 377 | 74 | |
GenBank mRNA | BG806503.1 | - | 2 - 3 | 302 | 74 |
CK618363.1 | - | 2 - 3 | 333 | 74 | |
CX206739.1 | - | 2 - 3 | 340 | 74 | |
CX220562.1 | - | 2 - 3 | 384 | 74 |