The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301176.1 | NP_001288105.1 | 13 - 14 | 1240 | 68 |
NM_025518.4 | NP_079794.1 | 14 - 15 | 1284 | 68 | |
GenBank mRNA | AK009391.1 | - | 14 - 15 | 1256 | 68 |
AK077451.1 | - | 13 - 14 | 1212 | 68 | |
BC058431.1 | - | 5 - 6 | 555 | 68 | |
BC116934.1 | - | 14 - 15 | 1202 | 68 | |
BC116936.1 | - | 14 - 15 | 1202 | 68 |