The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030176.2 | NP_084452.2 | 2 - 3 | 391 | 71 |
GenBank mRNA | AK019166.1 | - | 1 - 2 | 716 | 71 |
AK081234.1 | - | 2 - 3 | 391 | 71 | |
AK170267.1 | - | 1 - 2 | 691 | 71 | |
AK220503.1 | - | - | 691 | 71 | |
BC139310.1 | - | 1 - 2 | 326 | 71 | |
BC139311.1 | - | 1 - 2 | 326 | 71 |