The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164053.1 | NP_001157525.1 | 1 - 2 | 208 | 86 |
GenBank mRNA | BY240874.1 | - | 1 - 2 | 178 | 86 |