Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rltpr
Entrez Gene ID
234695
Gene Name
RGD motif, leucine rich repeats, tropomodulin domain and proline-rich containing
Gene Aliases
CG1399-PB, D130029J02Rik, Gm585
Chromosome Location
Chr.8: 105686010 - 105698195 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033320.2 NP_001028492.1 3 - 4 563 69
GenBank mRNA AK031012.1 - 3 - 4 562 69
HF678090.1 - 24 - 25 2194 69

View Full Product Details