The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039501.2 | NP_001034590.1 | 1 - 2 | 151 | 93 |
GenBank mRNA | AK131754.1 | - | 1 - 2 | 152 | 93 |
BC132270.1 | - | 1 - 2 | 142 | 93 | |
BC132272.1 | - | 1 - 2 | 142 | 93 | |
DQ437333.1 | - | 1 - 2 | 257 | 93 |