Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Wfdc3
- Entrez Gene ID
-
71856
- Gene Name
- WAP four-disulfide core domain 3
- Gene Aliases
-
1700015L13Rik, 1700127F16Rik
- Chromosome Location
-
Chr.2: 164731226 - 164745910 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK006001.1 |
- |
1 - 2 |
100 |
78 |
View Full Product Details