Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Sys1
- Entrez Gene ID
-
66460
- Gene Name
- SYS1 Golgi-localized integral membrane protein homolog (S. cerevisiae)
- Gene Aliases
-
2610042O14Rik, AI266964, C87389
- Chromosome Location
-
Chr.2: 164456981 - 164479637 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK032073.1 |
- |
3 - 4 |
424 |
79 |
View Full Product Details