Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Matn4
Entrez Gene ID
17183
Gene Name
matrilin 4
Gene Aliases
matrilin-4
Chromosome Location
Chr.2: 164389393 - 164405160 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252563.1 NP_001239492.1 10 - 11 1920 87
NM_013592.4 NP_038620.2 10 - 11 2185 87
GenBank mRNA AJ006140.1 - 10 - 11 1904 87
AK041073.1 - 10 - 11 1893 87
BC036558.1 - 10 - 11 1920 87

View Full Product Details