Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Matn4
Entrez Gene ID
17183
Gene Name
matrilin 4
Gene Aliases
matrilin-4
Chromosome Location
Chr.2: 164389393 - 164405160 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252563.1 NP_001239492.1 5 - 6 1002 81
NM_013592.4 NP_038620.2 5 - 6 1267 81
GenBank mRNA AJ006140.1 - 5 - 6 986 81
AK041073.1 - 5 - 6 975 81
AK160786.1 - 5 - 6 1063 81
BC036558.1 - 5 - 6 1002 81

View Full Product Details