Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Trp53rk
- Entrez Gene ID
-
76367
- Gene Name
- transformation related protein 53 regulating kinase
- Gene Aliases
-
2810408M09Rik, 4933401B08Rik, 5630401H01Rik, Nori-2, PRPK, Tp53rk, Trp53tk, mNori-2p
- Chromosome Location
-
Chr.2: 166792518 - 166799492 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK077074.1 |
- |
1 - 2 |
179 |
75 |
View Full Product Details