The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019835.2 | NP_062809.2 | 8 - 9 | 1164 | 62 |
GenBank mRNA | AB004786.1 | - | 8 - 9 | 1165 | 62 |
AF142673.1 | - | 8 - 9 | 1018 | 62 | |
AK151469.1 | - | 5 - 6 | 634 | 62 | |
AK152652.1 | - | 5 - 6 | 634 | 62 |