Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ptgis
Entrez Gene ID
19223
Gene Name
prostaglandin I2 (prostacyclin) synthase
Gene Aliases
Cyp8, Cyp8a1, Pgis
Chromosome Location
Chr.2: 167203194 - 167252127 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008968.3 NP_032994.1 7 - 8 1149 68
GenBank mRNA AB001607.1 - 7 - 8 1067 68
AK048059.1 - - 1202 68
AK076585.1 - 7 - 8 1150 68
BC062151.1 - 7 - 8 1156 68

View Full Product Details