Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ptgis
Entrez Gene ID
19223
Gene Name
prostaglandin I2 (prostacyclin) synthase
Gene Aliases
Cyp8, Cyp8a1, Pgis
Chromosome Location
Chr.2: 167203194 - 167252127 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK048059.1 - - 1526 121

View Full Product Details