Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sulf2
Entrez Gene ID
72043
Gene Name
sulfatase 2
Gene Aliases
2010004N24Rik, AU020235, MSulf-2, mKIAA1247
Chromosome Location
Chr.2: 166073899 - 166155683 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252578.1 NP_001239507.1 11 - 12 2028 73
NM_001252579.1 NP_001239508.1 11 - 12 1874 73
NM_028072.5 NP_082348.2 11 - 12 1870 73
GenBank mRNA AK034712.1 - 11 - 12 1855 73
AK036685.1 - 11 - 12 1806 73
AK049170.1 - 6 - 7 785 73
AK081643.1 - 11 - 12 1828 73
AK129316.1 - 11 - 12 2028 73
AK133336.1 - - 1844 73
AK165183.1 - 11 - 12 1782 73
AY101177.1 - 10 - 11 1689 73
BC062900.1 - 11 - 12 1826 73
BC141086.1 - 10 - 11 1623 73

View Full Product Details