Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Elmo2
Entrez Gene ID
140579
Gene Name
engulfment and cell motility 2
Gene Aliases
1190002F24Rik, CED-12
Chromosome Location
Chr.2: 165288031 - 165326479 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080287.2 NP_525026.2 12 - 13 1162 71
NM_207706.1 NP_997589.1 13 - 14 1198 71
GenBank mRNA AB093301.1 - 3 - 4 1743 71
AF398884.1 - 11 - 12 1061 71
AK032033.1 - 13 - 14 1171 71
AK033918.1 - 12 - 13 1356 71
AK038097.1 - 12 - 13 1151 71
AK038455.1 - 12 - 13 1268 71
AK045428.1 - 12 - 13 1163 71
AK047040.1 - 12 - 13 1124 71
AK050823.1 - 12 - 13 1125 71
AK053574.1 - 13 - 14 1162 71
AK129023.1 - 12 - 13 1162 71
BC023954.1 - 13 - 14 1183 71
BC033418.1 - 11 - 12 1092 71

View Full Product Details