Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfp64
Entrez Gene ID
22722
Gene Name
zinc finger protein 64
Gene Aliases
-
Chromosome Location
Chr.2: 168925361 - 168955587 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009564.2 NP_033590.2 3 - 4 868 71
GenBank mRNA AK010444.1 - 3 - 4 570 71
AK146036.1 - 3 - 4 554 71
BC004695.1 - - 550 71
CB182814.2 - 2 - 3 312 71
CX205658.1 - 3 - 4 543 71
U49046.1 - 3 - 4 540 71

View Full Product Details