Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Stx16
- Entrez Gene ID
-
228960
- Gene Name
- syntaxin 16
- Gene Aliases
-
4930401D03, 5430410K23Rik, 6330500A18Rik, AI648908, AW553605, Syn16
- Chromosome Location
-
Chr.2: 174076262 - 174099771 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK029571.1 |
- |
1 - 2 |
64 |
58 |
View Full Product Details