Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wnt9b
Entrez Gene ID
22412
Gene Name
wingless-type MMTV integration site family, member 9B
Gene Aliases
Wnt14b, Wnt15, clf, clf1, wnt-14b, wnt-15
Chromosome Location
Chr.11: 103727363 - 103749821 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011719.4 NP_035849.3 1 - 2 135 133
GenBank mRNA AB073819.1 - 1 - 2 98 133
AK052708.1 - 1 - 2 136 133
BC110482.1 - 1 - 2 127 133

View Full Product Details