The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282967.1 | NP_001269896.1 | 2 - 3 | 621 | 63 |
GenBank mRNA | FN434121.1 | - | 1 - 2 | 153 | 63 |