Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krr1
Entrez Gene ID
52705
Gene Name
KRR1, small subunit (SSU) processome component, homolog (yeast)
Gene Aliases
2610511F02Rik, AI255219, AI428520, D10Ertd773e, Hrb2
Chromosome Location
Chr.10: 111972695 - 111988430 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178610.4 NP_848725.2 3 - 4 486 59
GenBank mRNA AK011972.1 - 3 - 4 396 59
AK012108.1 - 3 - 4 424 59
AK030560.1 - 3 - 4 398 59
AK032472.1 - 3 - 4 398 59
AK036447.1 - 3 - 4 407 59
AK041854.1 - 3 - 4 398 59
AK146926.1 - 3 - 4 404 59
AK168675.1 - 3 - 4 398 59
BC089510.1 - 3 - 4 403 59
BC094236.1 - 3 - 4 387 59

View Full Product Details