The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037713.4 | NP_001032802.2 | 2 - 3 | 213 | 66 |
GenBank mRNA | AK140315.1 | - | 2 - 3 | 178 | 66 |
CK130151.1 | - | 1 - 2 | 148 | 66 |