Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pitpnm3
Entrez Gene ID
327958
Gene Name
PITPNM family member 3
Gene Aliases
A330068P14Rik, AI848332, Ackr6, Gm880
Chromosome Location
Chr.11: 72047528 - 72135889 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024927.2 NP_001020098.1 12 - 13 1841 70
NM_001081641.1 NP_001075110.1 11 - 12 1793 70
GenBank mRNA AK147444.1 - 12 - 13 1798 70
AK147630.1 - 11 - 12 1626 70

View Full Product Details