The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024927.2 | NP_001020098.1 | 1 - 2 | 240 | 88 |
NM_001081641.1 | NP_001075110.1 | 1 - 2 | 240 | 88 | |
GenBank mRNA | AK039594.1 | - | 1 - 2 | 129 | 88 |
AK147444.1 | - | 1 - 2 | 197 | 88 | |
AK147630.1 | - | 1 - 2 | 73 | 88 | |
CJ123564.1 | - | 1 - 2 | 241 | 88 |