Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tsr1
Entrez Gene ID
104662
Gene Name
TSR1 20S rRNA accumulation
Gene Aliases
AU040765, AW550801, mKIAA1401
Chromosome Location
Chr.11: 74898080 - 74909340 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177325.3 NP_796299.2 4 - 5 589 82
GenBank mRNA AK030545.1 - 3 - 4 495 82
AK129350.1 - 4 - 5 543 82
AK134365.1 - 4 - 5 573 82
AK154063.1 - 4 - 5 576 82
AK165662.1 - 4 - 5 580 82
BC031531.1 - 4 - 5 562 82

View Full Product Details