Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sirt7
Entrez Gene ID
209011
Gene Name
sirtuin 7
Gene Aliases
-
Chromosome Location
Chr.11: 120618372 - 120625002 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153056.2 NP_694696.2 4 - 5 435 62
GenBank mRNA AK135484.1 - 4 - 5 432 62
AY251540.1 - 4 - 5 411 62
BC026403.1 - 4 - 5 400 62
BC026637.1 - 4 - 5 403 62
BC026650.1 - 4 - 5 403 62
BC040759.1 - 4 - 5 400 62
BC080304.1 - 3 - 4 739 62

View Full Product Details