Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ces2d-ps
- Entrez Gene ID
-
667754
- Gene Name
- carboxylesterase 2D, pseudogene
- Gene Aliases
-
ENSMUSG00000031884, Gm9756
- Chromosome Location
-
Chr.8: 104867424 - 104874083 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_033726.1 |
|
2 - 3 |
348 |
72 |
View Full Product Details