Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Hexdc
- Entrez Gene ID
-
238023
- Gene Name
- hexosaminidase (glycosyl hydrolase family 20, catalytic domain) containing
- Gene Aliases
-
BC069960
- Chromosome Location
-
Chr.11: 121204433 - 121222656 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC056487.1 |
- |
5 - 6 |
2493 |
61 |
View Full Product Details