The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001112699.1 | NP_001106169.1 | 2 - 3 | 237 | 87 |
NM_001112700.1 | NP_001106170.1 | 2 - 3 | 177 | 87 | |
NM_011180.3 | NP_035310.2 | 2 - 3 | 237 | 87 | |
GenBank mRNA | AB013464.1 | - | 2 - 3 | 158 | 87 |
AB035538.1 | - | 2 - 3 | 206 | 87 | |
AF051337.1 | - | 1 - 2 | 105 | 87 | |
AK078502.1 | - | - | 148 | 87 | |
AK158216.1 | - | 2 - 3 | 177 | 87 | |
AK172296.1 | - | 2 - 3 | 182 | 87 | |
AK220302.1 | - | 2 - 3 | 159 | 87 | |
AY126516.1 | - | 2 - 3 | 206 | 87 | |
BC057974.1 | - | 2 - 3 | 121 | 87 | |
BY204859.1 | - | 2 - 3 | 177 | 87 | |
CJ131185.1 | - | 2 - 3 | 237 | 87 |