Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyth1
Entrez Gene ID
19157
Gene Name
cytohesin 1
Gene Aliases
CLM1, CTH-1, CYTIP, Pscd1, mKIAA4240
Chromosome Location
Chr.11: 118164166 - 118248592 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112699.1 NP_001106169.1 4 - 5 369 74
NM_001112700.1 NP_001106170.1 4 - 5 309 74
NM_011180.3 NP_035310.2 4 - 5 369 74
GenBank mRNA AB013464.1 - 4 - 5 290 74
AB035538.1 - 4 - 5 338 74
AF051337.1 - 3 - 4 237 74
AK078502.1 - - 280 74
AK158216.1 - 4 - 5 309 74
AK172296.1 - 4 - 5 314 74
AK220302.1 - 4 - 5 291 74
AY126516.1 - 4 - 5 338 74
BC057974.1 - 4 - 5 253 74
CJ131185.1 - 4 - 5 369 74

View Full Product Details