The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133660.3 | NP_598421.1 | 10 - 11 | 1360 | 66 |
GenBank mRNA | AK144549.1 | - | 10 - 11 | 1345 | 66 |
BC019208.1 | - | 10 - 11 | 1291 | 66 | |
S80191.1 | - | 10 - 11 | 1290 | 66 |