The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144930.2 | NP_659179.1 | 1 - 2 | 93 | 73 |
GenBank mRNA | AK136491.1 | - | 1 - 2 | 85 | 73 |
BC013479.1 | - | 1 - 2 | 68 | 73 |